ContactUsLinkComponent
REAGENTS HIGH SENSITIVITY D5000
High Sensitivity DNA ScreenTape Analysis
Catalog # CA76645-492
undefined
High Sensitivity DNA ScreenTape Analysis
Catalog # CA76645-492
Supplier Number:  5067-5593
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    High sensitivity D5000 reagents
  • Cat. no.:
    CA76645-492
  • Details:
    For the high sensitivity analysis of DNA from 100 to 5000 bp, includes ladder and sample buffer sufficient for 105 samples, order with 5067-5592.
Specifications
About this item
DNA fragment or library analysis from samples of limited abundance, for samples up to 5000 bp.
  • The broad size range allows analysis of DNA fragments or libraries from 35 to 5000 bp
  • Results are obtained quickly, in approximately 1 minute per sample
  • Detection of DNA samples with limited abundance is possible, for samples down to 5 pg/µl
  • Ready-to use ScreenTape devices, pre-packaged reagent and automated data analysis offer an easy to use system with reproducible DNA sizing and quantification results
  • Reliable sample quality control is provided for next-generation sequencing (NGS) workflows
  • No cross contamination or sample carryover is caused due to the use of single lanes and individual loading tips
  • Partially used ScreenTape devices can be used to completion at a later point in time
Agilent's TapeStation High Sensitivity DNA system provides a fast, automated, and flexible solution for samples of limited abundance in many applications. High Sensitivity DNA electrophoresis can be used for quality control of next-generation sequencing (NGS) libraries.
The High Sensitivity DNA ScreenTape assays are suitable for analysis of precious samples. They can separate DNA fragments and libraries up to 5000 base pairs in 1 to 2 minutes per sample. Data output yields objective quantitative and purity measurements from only 2 µl of sample.
Assays are compatible with all Agilent TapeStation instruments.
Note: Some products within these assays contain SYBR™ Green I, which is licensed from Life Technologies Corporation for use in research and development only. SYBR™ is a registered trademark of Life Technologies Corporation.
Caution: For Research Use Only. Not for use in diagnostic procedures.
Frequently Bought Together