Order Entry
Canada
Orders LinkContactUsLinkComponent
Fluid A, USP

Fluid A, USP

Fluid A, USP, 1 L, 1 L, Bottle, —
Catalog # CA89407-526
Supplier #:  U208
Fluid A, USP
Catalog # CA89407-526
CAS Number:  
Fluid A, USP
Fluid A, USP

Fluid A, USP, 1 L, 1 L, Bottle, —

CAS Number:  
Price
$923.00
Log in for availability
Pack of 10
Minimum order quantity is 1
Product Details & Documents

A rinsing and diluting fluid for use in USP testing protocols.

A rinsing and diluting fluid for use in USP testing protocols. Contains peptone 0.1%.

Hardy Diagnostics' sterility media and rinse solutions are manufactured in an ISO 13485 environmentally controlled production center. Each lot is certified for pH, sterility and growth promotion using ATCC® strains specified by the USP. Our approach to manufacturing ensures all sterility media and rinse fluids provide the highest level of clarity for improved reading accuracy, thus significantly reducing the risk of incorrect interpretation and false results.

Ordering information: Order by the pack of 10

Specifications

Specifications

  • Pack type:
    Bottle
  • Description:
    Fluid A, USP
  • Size:
    1 L
  • Minimum Order Quantity:
  • Volume:
    1 L

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.