Order Entry
Canada
Orders LinkContactUsLinkComponent
Wearable Circuits

Wearable Circuits

Wearable circuits, classroom pack
Catalog # 470348-934
Supplier:   BROWN DOG GADGETS
Supplier #:  K1060-CLS
Wearable Circuits
Catalog # 470348-934
Supplier:  BROWN DOG GADGETS
Wearable Circuits
Wearable Circuits

Wearable circuits, classroom pack

Price
181,99 $
Log in for availability
Each
Product Details & Documents

Introduce core science and engineering concepts within an arts context.

  • Engage makers of all ages
  • Put the art in STEAM activities - learn about circuits while crafting
  • No additional tools needed, not recommended for sewing machines

Engage learners of all ages by adding circuity to hand-crafted wearable items. Wearable Circuits kits make this idea approachable for all learners regardless of dexterity issues, classroom time constraints, or existing sewing skills. Use our conductive Maker Tape™ and raw circuit components to add a variety of LED accents to virtually any wearable item that you can stick a piece of tape to.

Not recommended for sewing machines. Fabric not included.

Delivery information: Standard pack includes (15) Jumbo LEDs, (5) CR2032 batteries, (5) Adhesive hook and loop dots, (1) 2" Conductive hook and loop adhesive strip, (2) Rolls of conductive Maker tape™, ¼" × 5 m, Downloadable instruction manual. Classroom pack includes (75) Jumbo LEDs; (25) CR2032 batteries; (25) Adhesive hook and loop dots, (1) 8" Conductive hook and loop adhesive strip; (2) Rolls of conductive Maker Tape™, ¼" × 20 m; Downloadable instruction manual.

Specifications
  • Description:
    Wearable circuits, classroom pack
Accessories

Product Information

  • Catalog #
    Availability
    Price
  • Conductive Maker Tape™, ¼" × 20 m
    Conductive Maker Tape™, ¼" × 20 m
    470234-666
    Log in for availability
    Each - $16.45

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.