Electro-Gel™ Electrolyte Replenishers
1 oz., 16 cups/pack
CA76628-300
: Bio-Serv
: EGS16P-1
Gelled, oral electrolyte hydration supplement. Provides balanced electrolyte and glucose levels to provide energy, nutrients and hydration to accelerate recovery.
- Economical and easy to use requiring no refrigeration or special storage
- Highly palatable with a sweet, salty orange or strawberry flavor
- Comparable to Pedialyte and PRANG in gel form
- Suitable for a variety of research species
- Thermoreversible
- Gamma Irradiated
Shelf life of 12 months, cool dry storage.
Ingredients: Purified water, dextrose, gelatin, L-ascorbic acid phosphate, sodium chloride, sodium bicarbonate, citric acid, potassium chloride, flavoring, natural color, potassium sorbate, acesulfame potassium.
- Size:1 oz.
- Packaging:16 cups/pack