Order Entry
Canada
Orders LinkContactUsLinkComponent
VWR® DNA Loading Buffers

VWR® DNA Loading Buffers

5X Loading Buffer Blue, 5× 1 ml
Catalog # 76620-552
Supplier:   VWR International
VWR® DNA Loading Buffers
Catalog # 76620-552
Supplier:  VWR International
CAS Number:  
VWR® DNA Loading Buffers
VWR® DNA Loading Buffers

5X Loading Buffer Blue, 5× 1 ml

CAS Number:  
Price
$72.98
Log in for availability
Each
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

VWR® loading buffers, with three different dyes and migration fronts, make it easy to select the right loading buffer for your specific task.

  • Ready-to-use buffers
  • 5x formulation
  • Three different tracking dyes available

VWR® loading buffers are formulated as 5x solutions containing Ficoll, Tris-buffer, EDTA and either Cresol Red, Bromophenol Blue or Orange G as tracking dye.

DNA loading buffers are used for loading DNA samples onto DNA gels for gel electrophoresis. DNA loading buffers contain a coloured dye and a density agent. The density agent serves to enhance the density of the DNA sample allowing the DNA to sink into the bottom of the well. The dye adds visibility to the DNA sample and also serves as a tracking dye allowing the user to monitor the DNA migration during electrophoresis.

For long term storage, keep the loading buffer at -20 °C.

Caution: For Research Use Only. Not for use in diagnostic procedures.

Specifications

Specifications

  • Description:
    5X Loading Buffer Blue
  • Volume:
    5× 1 ml

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.