Order Entry
Canada
Orders LinkContactUsLinkComponent
VWR® dNTPs

VWR® dNTPs

dNTP Set (100mM of each dATP, dCTP, dGTP, dTTP), 100 μmoL of each, 16× 250 µl, EA
Catalog # 76620-548
Supplier:   VWR International
VWR® dNTPs
Catalog # 76620-548
Supplier:  VWR International
CAS Number:  
VWR® dNTPs
VWR® dNTPs

dNTP Set (100mM of each dATP, dCTP, dGTP, dTTP), 100 μmoL of each, 16× 250 µl, EA

CAS Number:  
Price
$1,035.63
Log in for availability
Each
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Ready to use molecular biology grade dNTP mixes and dNTP sets.

  • Available as pre-mixed 40 or 100 mM solutions , or as sets of individual 100 mM dNTP solutions
  • Both pre-mixed and sets have been functionally tested in PCR
  • Purity >99% by HPLC
  • Supplied in solution at pH 7.3 to 7.5

The dNTP mix is designed to save hands-on time for researchers, and reduce the possibility of contamination by reducing pipetting. The dNTP mix and dNTP solutions are convenient for use in DNA polymerization reactions, DNA labelling and sequencing processes.

Caution: For Research Use Only. Not for use in diagnostic procedures.

Specifications

Specifications

  • Description:
    dNTP Set (100mM of each dATP, dCTP, dGTP, dTTP)
  • Size:
    100 μmoL of each
  • Units:
    EA
  • Volume:
    16× 250 µl

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.