Orders LinkContactUsLinkComponent
Large Pathology Containers, Mopec
Large Pathology Containers, Mopec
3.5 gal., Large pathology containers
Catalog # 76579-434
Supplier:   Mopec
Supplier #:  BG635
Large Pathology Containers, Mopec
Catalog # 76579-434
Supplier:  Mopec
Large Pathology Containers, Mopec
Large Pathology Containers, Mopec
3.5 gal., Large pathology containers
Price
$159.43
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Mopec’s screw top pathology containers are made from a high-quality, clarified polypropylene material. The screw-top lid creates a moisture and tamper-resistant closure that increases safety for laboratory employees and helps protect pathology specimens during transportation. Features an attached handle for easy transportation. Available in multiple sizes.
  • Made from a high-quality, clarified polypropylene material
  • Screw-top lid creates a moisture and tamper-resistant closure
  • Features an attached handle for easy transportation
Sold individually.
Specifications
  • Capacity:
    3.5 gal.
  • Description:
    Large pathology containers
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)