Eisco High-Performance Variable Volume Micropipettes
2 - 20 µl, 0.10 µl, Red, 1
Catalog # 470345-790
Supplier: Wards
Supplier #: VVM8
Jump to:
Product Details & Documents
Variable volume micropipettes are fully autoclavable and feature a simple ergonomic design for comfort, color coded plungers, and calibration conforming to DIN ISO standards.
- Soft and lightweight operation with an ergonomic, soft touch tip ejector designed to minimize hand fatigue, reduce thumb stress and to increase comfort with prolonged use
- Fully autoclavable with a durable, autoclavable PVDF tip cone
- Slip-resistant thermoplastic elastomer (TPE) body grip to minimize body heat transfer to the micropipette during long use, resulting in higher accuracy
- Color coded plungers for easy identification
- Calibration conforms to DIN 12650 & EN-ISO 8655 standard ensuring high accuracy and precision - Calibration report enclosed with every micropipette
High performance variable volume micropipettes have a simple design which allows for easy adjustment and reading of increments.
Specifications
- Volume:2 - 20 µl
- Increments:0.10 µl
- No. of channels:1
- Color code:Red