Orders LinkContactUsLinkComponent
Eisco Rectangular Retort Bases, Cast Iron
8×5", 3 lbs.
Catalog # 470345-054
Supplier:   Wards
 :  CH0653B
Eisco Rectangular Retort Bases, Cast Iron
Catalog # 470345-054
Supplier:  Wards
Eisco Rectangular Retort Bases, Cast Iron
Eisco Rectangular Retort Bases, Cast Iron
8×5", 3 lbs.
Price
$26.45
Log in for availability
Each
Product Details & Documents
Heavy Retort Bases with tapped rod hole made of chemically-resistant coated cast iron.
  • Rectangular retort bases made of chemical-resistant cast iron with coated finished
  • Heavy and sturdy construction provides superb stability for experimental setups
  • Features a tapped hole for a 10×1.5 mm threaded rod (not included)
Heavy-weighted Retort Stands are made of chemically-resistant cast iron with a coated finish, and feature a tapped hole at one end, suitable for rods with a 10×1.5 mm metric thread. Provides superb stability for basic experiments and setups. The durable and sturdy construction of these bases make them an excellent choice for science classrooms.
Specifications
  • Base dimensions:
    8×5"
  • Weight:
    3 lbs.
Accessories
Product Information
  • Catalog #
    Ø
    Rod Length
    Availability
    Price
  • Aluminum support rod
    Aluminum support rod
    470345-328
    12 mm
    1016 mm (40")
    Log in for availability
    Each - $31.35
Product Information
  • Catalog #
    Ø
    Rod Length
    Availability
    Price
  • Aluminum support rod
    Aluminum support rod
    470345-052
    12 mm
    762 mm (30")
    Log in for availability
    Each - $25.40
Product Information
  • Catalog #
    Ø
    Rod Length
    Availability
    Price
  • Steel support rod
    Steel support rod
    470345-310
    12 mm
    508 mm (20")
    Log in for availability
    Each - $17.00
Product Information
  • Catalog #
    Ø
    Rod Length
    Availability
    Price
  • Stainless steel support rod
    Stainless steel support rod
    470344-218
    12 mm
    609 mm (24")
    Log in for availability
    Each - $24.25
Product Information
  • Catalog #
    Ø
    Rod Length
    Availability
    Price
  • Stainless steel support rod
    Stainless steel support rod
    470344-962
    12 mm
    901 mm (35.5")
    Log in for availability
    Each - $38.25
Product Information
  • Catalog #
    Ø
    Rod Length
    Availability
    Price
  • Steel support rod
    Steel support rod
    470345-394
    12 mm
    609 mm (24")
    Log in for availability
    Each - $22.65
Product Information
  • Catalog #
    Ø
    Rod Length
    Availability
    Price
  • Aluminum support rod
    Aluminum support rod
    470343-698
    12 mm
    609 mm (24")
    Log in for availability
    Each - $19.10
Product Information
  • Catalog #
    Ø
    Rod Length
    Availability
    Price
  • Stainless steel support rod
    Stainless steel support rod
    470344-578
    12 mm
    762 mm (30")
    Log in for availability
    Each - $28.95
Product Information
  • Catalog #
    Ø
    Rod Length
    Availability
    Price
  • Stainless steel support rod
    Stainless steel support rod
    470345-364
    12 mm
    508 mm (20")
    Log in for availability
    Each - $22.65
Product Information
  • Catalog #
    Ø
    Rod Length
    Availability
    Price
  • Steel support rod
    Steel support rod
    470345-166
    12 mm
    762 mm (30")
    Log in for availability
    Each - $18.40
Product Information
  • Catalog #
    Ø
    Rod Length
    Availability
    Price
  • Aluminum support rod
    Aluminum support rod
    470345-558
    12 mm
    508 mm (20")
    Log in for availability
    Each - $17.05
Product Information
  • Catalog #
    Ø
    Rod Length
    Availability
    Price
  • Stainless steel support rod
    Stainless steel support rod
    470345-774
    12 mm
    1016 mm (40")
    Log in for availability
    Each - $38.25