Orders LinkContactUsLinkComponent
Eisco® Hooked Weights, Iron
Eisco® Hooked Weights, Iron
500 g
Catalog # 470344-630
Supplier:   Wards
Supplier #:  PHHWFE1
Eisco® Hooked Weights, Iron
Catalog # 470344-630
Supplier:  Wards
Eisco® Hooked Weights, Iron
Eisco® Hooked Weights, Iron
500 g
Price
$31.10
Log in for availability
Each
Product Details & Documents
Hooked Iron Weights with cylindrical bodies and weights printed on each piece for easy identification.
  • Made from high-grade iron
  • Integral hook for easy lifting and hanging
  • Weight printed on each piece for easy identification
  • Ideal as additional or spare part for hooked weight sets
Cylindrical single iron weights feature a hook for easy lifting and hanging. Weight value is printed on each piece for easy identification. Great as replacements for missing pieces or as spares.
Specifications
  • Weight:
    500 g