Orders LinkContactUsLinkComponent
Eisco Plastic Scoops
Plastic scoops, 100 ml
  470343-970
Eisco Plastic Scoops
  470343-970
 :  
Eisco Plastic Scoops
Eisco Plastic Scoops
Plastic scoops, 100 ml
Pack of 12 or 6 weighing scoops with labeled handles and flat bottoms.
  • Made of heat- and chemical-resistant polypropylene plastic
  • Scoop handles are labeled with capacity for easy identification
  • Flat bottom design allows scoops to rest upright for weighing and filling
Autoclavable and dishwasher safe for easy cleaning.
  • Capacity:
    100 ml
  • Description:
    Plastic scoops