Eisco® Hooked Weights, Iron
1000 g
Catalog # 470343-904
Supplier: Wards
Supplier #: PHHWFE2
Jump to:
Product Details & Documents
Hooked Iron Weights with cylindrical bodies and weights printed on each piece for easy identification.
- Made from high-grade iron
- Integral hook for easy lifting and hanging
- Weight printed on each piece for easy identification
- Ideal as additional or spare part for hooked weight sets
Cylindrical single iron weights feature a hook for easy lifting and hanging. Weight value is printed on each piece for easy identification. Great as replacements for missing pieces or as spares.
Specifications
- Weight:1000 g