Eisco Rectangular Retort Bases, Cast Iron
10×6", 4.6 lbs.
Catalog # 470343-652
Supplier: Wards
Supplier #: CH0653C
Jump to:
Product Details & Documents
Heavy Retort Bases with tapped rod hole made of chemically-resistant coated cast iron.
- Rectangular retort bases made of chemical-resistant cast iron with coated finished
- Heavy and sturdy construction provides superb stability for experimental setups
- Features a tapped hole for a 10×1.5 mm threaded rod (not included)
Heavy-weighted Retort Stands are made of chemically-resistant cast iron with a coated finish, and feature a tapped hole at one end, suitable for rods with a 10×1.5 mm metric thread. Provides superb stability for basic experiments and setups. The durable and sturdy construction of these bases make them an excellent choice for science classrooms.
Specifications
- Base dimensions:10×6"
- Weight:4.6 lbs.
Accessories
Product Information
- Catalog #ØRod LengthAvailabilityPrice
- Aluminum support rod
Product Information
- Catalog #ØRod LengthAvailabilityPrice
- Aluminum support rod
Product Information
- Catalog #ØRod LengthAvailabilityPrice
- Steel support rod
Product Information
- Catalog #ØRod LengthAvailabilityPrice
- Stainless steel support rod
Product Information
- Catalog #ØRod LengthAvailabilityPrice
- Stainless steel support rod
Product Information
- Catalog #ØRod LengthAvailabilityPrice
- Steel support rod
Product Information
- Catalog #ØRod LengthAvailabilityPrice
- Aluminum support rod
Product Information
- Catalog #ØRod LengthAvailabilityPrice
- Stainless steel support rod
Product Information
- Catalog #ØRod LengthAvailabilityPrice
- Stainless steel support rod
Product Information
- Catalog #ØRod LengthAvailabilityPrice
- Steel support rod
Product Information
- Catalog #ØRod LengthAvailabilityPrice
- Aluminum support rod
Product Information
- Catalog #ØRod LengthAvailabilityPrice
- Stainless steel support rod



