$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Canada
ContactUsLinkComponent
PALLADIUM 10 UG/ML (10 PPM) 30ML
SXCLPD1-1AM
:
SPEX CERTIPREP LLC
PALLADIUM 10 UG/ML (10 PPM) 30ML
SXCLPD1-1AM
:
SPEX CERTIPREP LLC
:
CLPD1-1AM
Frequently Bought Together
pH Reference Standard Buffers, VWR Chemicals BDH®
: BDH
VWR® Light-Duty Tissue Wipers
: VWR International
VWR® Universal Pipette Tips
: VWR International
VWR® Microcentrifuge Tubes, PP
: VWR International
Falcon® Centrifuge Tubes, Conical Bottom, Corning
: Corning
VWR® Nitrile Examination Gloves, Blue
: VWR International
VWR® Disposable Petri Dishes
: VWR International