ContactUsLinkComponent
Masterflex® Cleanroom ¾" Sanitary Tri-Clamp × ½" Barb Adapter in PVDF; 10 pack
Masterflex® Cleanroom ¾" Sanitary Tri-Clamp × ½" Barb Adapter in PVDF; 10 pack
Catalog # MFLX40330-16
Supplier:  VWR International
undefined
Masterflex® Cleanroom ¾" Sanitary Tri-Clamp × ½" Barb Adapter in PVDF; 10 pack
Catalog # MFLX40330-16
Supplier:  VWR International
About this item
Maintain sanitary conditions with secure connections from sanitary Tri-Clamp to tubing.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Meet requirements for USP Class VI, FDA, REACH, and California Proposition 65
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Packaged to exacting tolerances under ISO Class 7 cleanroom conditions
  • Compatible with other Tri-Clamp® connections
The sanitary Tri-Clamp barbed adapters are ideal for use in the biopharmaceutical, food and beverage, and single-use industries. They have excellent chemical resistance and may be sterilized by gamma irradiation, EtO, E-beam, or autoclave. The PVDF fittings withstand higher temperatures than the Polypropylene fittings. Clamps and gaskets required for a complete system are sold separately.
  • Barb size/tubing int.Ø (port 2):
    ½"
  • Connection Type:
    Sanitary to Barb
  • Flow pattern:
    Straight
  • Sanitary clamp size (Port 1):
    ¾"
  • Description:
    Cleanroom ¾" sanitary tri-clamp × ½" barb adapter in PVDF; 10 pack
  • Environmentally Preferable:
  • No. of ports:
    2
  • Regulatory approvals:
    FDA;USP
  • Sterilisation:
    Gamma irradiation, EtO, autoclave
  • Cat. no.:
    MFLX40330-16
  • Fitting Material:
    PVDF
  • Sanitary Gasket Size:
    ¾"
Frequently Bought Together