ContactUsLinkComponent
Masterflex® Fitting, PVDF, Straight, Hose Barb Plug, 1/4" ID; 10/PK
Masterflex® Fitting, PVDF, Straight, Hose Barb Plug, 1/4" ID; 10/PK
Catalog # MFLX40615-43
Supplier:  VWR International
undefined
Masterflex® Fitting, PVDF, Straight, Hose Barb Plug, 1/4" ID; 10/PK
Catalog # MFLX40615-43
Supplier:  VWR International
About this item
Protect against contaminants in tubing lines.
  • Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Use these barbed press-in-plugs to keep open tubing lines or tubing assemblies free of foreign contaminants and ensure process purity. Plugs are manufactured from 100% virgin materials in a ISO Class 8 (formerly Class 100,000) environment and include full material traceability. Available in PVDF or nylon. All plugs are sterilizable by gamma irradiation; nylon plugs may also be autoclaved.
  • Barb size/tubing int.Ø (port 1):
    ¼"
  • Connection Type:
    Barbed Plug
  • Flow pattern:
    Straight
  • Description:
    Fitting, PVDF, Straight, Hose Barb Plug, ¼" ID; 10/PK
  • Environmentally Preferable:
  • Cat. no.:
    MFLX40615-43
  • Fitting Material:
    PVDF
Frequently Bought Together