InfinityLab PlusPore, Agilent Technologies
Guard column, OligoPore, 6 µm, 7.50 mm, 50 mm, 0 - 3300 Da, Mixed Pore Size
Catalog # CAAGPL11131320
Supplier #: PL1113-1320
Jump to:
Discontinued
Product Details & Documents
PlusPore GPC columns offer the highest resolution gel permeation chromatography.
- PlusPore GPC columns offer a major resolution improvement over traditional mixed medias and single pore columns
- High-efficiency small particles use multipore technology, maximizing resolution without sacrificing molecular weight range or lifetime
- OligoPore uses unique ultrahigh pore volume media to achieve superior separation of polymers below 3,300 Daltons
PLgel individual pore size GPC columns offer the highest resolution within a narrow molecular weight range. Columns may be stacked to extend the molecular weight range. Individual pore columns are generally recommended for very small and very large polymers, which can be difficult to resolve using more common PLgel MIXED columns.
Specifications
- Column Format:Guard column
- Ø int.:7.50 mm
- Particle Size:6 µm
- Length:50 mm
- Packing material:Mixed Pore Size
- Phase:OligoPore
- Exclusion limit:0 - 3300 Da
- Cat. No.:CAAGPL11131320
- Max. pressure:150 bar
- Temperature limit:110 °C
- pH range:1-14



