Orders LinkContactUsLinkComponent
Disintegration Baskets, Agilent Technologies
Basket, 1-tube, 44 mm OD, 10-mesh, 1905 µm
Catalog # CAAG37-3040
 :  37-3040
Disintegration Baskets, Agilent Technologies
Catalog # CAAG37-3040
Supplier:  
Disintegration Baskets, Agilent Technologies
Disintegration Baskets, Agilent Technologies
Basket, 1-tube, 44 mm OD, 10-mesh, 1905 µm
Discontinued
Product Details & Documents
Agilent offers disintegration baskets for the 100 automated disintegration apparatus.
Specifications
  • Description:
    Basket, 1-tube, 44 mm OD, 10-mesh, 1905 µm