Orders LinkContactUsLinkComponent
Disintegration Baskets, Agilent Technologies
Disintegration Baskets, Agilent Technologies
Basket, 6-tube, 20-mesh, 864 µm
Catalog # CAAG37-3010
Supplier #:  37-3010
Disintegration Baskets, Agilent Technologies
Catalog # CAAG37-3010
Disintegration Baskets, Agilent Technologies
Disintegration Baskets, Agilent Technologies
Basket, 6-tube, 20-mesh, 864 µm
Price
$973.00
Log in for availability
Each
Product Details & Documents
Agilent offers disintegration baskets for the 100 automated disintegration apparatus.
Specifications
  • Description:
    Basket, 6-tube, 20-mesh, 864 µm