Dissolution Verification Tools, Agilent Technologies
Verified Height tool, stainless steel, sets height of basket and paddles, used for testing antibiotics, 45 mm, with certificate, for 708-DS
Catalog # CAAG12-7330V
Jump to:
Discontinued
Product Details & Documents
A variety of different height gauges and spacers to meet all needed regulatory specifications.
- Available for the Agilent 708-DS and legacy instruments
Verification and calibration tools can help you to install your accessories properly. Agilent provides a variety of different height gauges and spacers to meet all needed regulatory specifications, such as mechanical qualification and physical parameter verification, for your dissolution test.
Specifications
- Description:Verified Height tool, stainless steel, sets height of basket and paddles, used for testing antibiotics, 45 mm, with certificate, for 708-DS
- Certification:Yes