Dissolution Verification Tools, Agilent Technologies
Height spacer, for attachment to basket shaft, 25 mm above bottom of vessel, for 708-DS
Catalog # CAAG12-7280
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
Supplier #: 12-7280
Dissolution Verification Tools, Agilent Technologies
Catalog # CAAG12-7280
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
Jump to:
Product Details & Documents
A variety of different height gauges and spacers to meet all needed regulatory specifications.
- Available for the Agilent 708-DS and legacy instruments
Verification and calibration tools can help you to install your accessories properly. Agilent provides a variety of different height gauges and spacers to meet all needed regulatory specifications, such as mechanical qualification and physical parameter verification, for your dissolution test.
Specifications
- Description:Height spacer, for attachment to basket shaft, 25 mm above bottom of vessel, for 708-DS
- Certification:No