Liston Bone Cutting Forceps, Angled, Mortech
Product Details & Documents
Effective bone cutting instrument for use during autopsies and post-mortem dissection. Sharp tips perform a clean extraction.
- Stainless steel
- Smooth grip
Available in a variety of lengths.
Specifications
- Length:7.5"
- Curvature:Angled
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)