Orders LinkContactUsLinkComponent
Liston Bone Cutting Forceps, Angled, Mortech
Liston Bone Cutting Forceps, Angled, Mortech
5.5", Angled
Catalog # 76549-246
Supplier:   Mopec
Supplier #:  AG052
Liston Bone Cutting Forceps, Angled, Mortech
Catalog # 76549-246
Supplier:  Mopec
Liston Bone Cutting Forceps, Angled, Mortech
Liston Bone Cutting Forceps, Angled, Mortech
5.5", Angled
Price
$157.02
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Effective bone cutting instrument for use during autopsies and post-mortem dissection. Sharp tips perform a clean extraction.
  • Stainless steel
  • Smooth grip
Available in a variety of lengths.
Specifications
  • Length:
    5.5"
  • Curvature:
    Angled
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)