Orders LinkContactUsLinkComponent
Forceps, Specimen Paddle, Mortech
Forceps, Specimen Paddle, Mortech
6", Round
Catalog # 76548-746
Supplier:   Mopec
Supplier #:  AB064
Forceps, Specimen Paddle, Mortech
Catalog # 76548-746
Supplier:  Mopec
Forceps, Specimen Paddle, Mortech
Forceps, Specimen Paddle, Mortech
6", Round
Price
$191.97
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Paddle forceps hold all types of tissue with the secure grip needed to accomplish thin sections for analysis. Thin sections are essential for proper infiltration of reagents prior to study. Great for training residents and technicians on thin section technique, cutting thin sections of fatty tissue like breast tissue and cutting even thickness sections of any type tissue.
  • Standard grade stainless steel
  • Ribbed grip
  • Paddle ends
Available in a variety of paddle types.
Specifications
  • Length:
    6"
  • Tip style:
    Round
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)