Orders LinkContactUsLinkComponent
Forceps, Ergonomic, Mortech
Forceps, Ergonomic, Mortech
5.5", Serrated
Catalog # 76548-736
Supplier:   Mopec
Supplier #:  AB058
Forceps, Ergonomic, Mortech
Catalog # 76548-736
Supplier:  Mopec
Forceps, Ergonomic, Mortech
Forceps, Ergonomic, Mortech
5.5", Serrated
Price
$191.97
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Unique perforated paddle handles for secure slip-free grip. Fingertip aligned perforations offer precise maneuvering. Extremely lightweight and versatile for a wide variety of tissue specimens. Can be cleaned with any laboratory cleaner or disinfectant.
  • Stainless steel
  • Broad grip
Available with smooth or serrated tips.
Specifications
  • Length:
    5.5"
  • Tip style:
    Serrated
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.