Orders LinkContactUsLinkComponent
Forceps, Russian Tissue, Mortech
Standard, 8"
Catalog # 76548-682
Supplier:   Mopec
 :  AB030
Forceps, Russian Tissue, Mortech
Catalog # 76548-682
Supplier:  Mopec
Forceps, Russian Tissue, Mortech
Forceps, Russian Tissue, Mortech
Standard, 8"
Price
$79.68
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Rounded perimeter multi-tooth tip allows for maximum tissue grip. Variety of lengths and grades.
  • Stainless steel
  • Ribbed grip
  • Rounded multi-toothed tips
Available in standard or premium grade.
Specifications
  • Length:
    8"
  • Grade:
    Standard
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.