Forceps, Russian Tissue, Mortech
Product Details & Documents
Rounded perimeter multi-tooth tip allows for maximum tissue grip. Variety of lengths and grades.
- Stainless steel
- Ribbed grip
- Rounded multi-toothed tips
Available in standard or premium grade.
Specifications
- Length:6"
- Grade:Standard
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)