Orders LinkContactUsLinkComponent
Forceps, Dissection, Serrated Tip, Mortech
Forceps, Dissection, Serrated Tip, Mortech
Standard, 4.5"
Catalog # 76548-626
Supplier:   Mopec
Supplier #:  AB001
Forceps, Dissection, Serrated Tip, Mortech
Catalog # 76548-626
Supplier:  Mopec
Forceps, Dissection, Serrated Tip, Mortech
Forceps, Dissection, Serrated Tip, Mortech
Standard, 4.5"
Price
$26.96
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Standard, high-quality, serrated forceps are available in a wide variety of lengths and grades.
  • Stainless steel
  • Ribbed grip
  • Serrated tips
Available in standard or premium grade.
Specifications
  • Length:
    4.5"
  • Grade:
    Standard
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)