Orders LinkContactUsLinkComponent
Scissors, Titanium, Iris, Mortech
Scissors, Titanium, Iris, Mortech
4", Straight
Catalog # 76548-622
Supplier:   Mopec
Supplier #:  AA707
Scissors, Titanium, Iris, Mortech
Catalog # 76548-622
Supplier:  Mopec
Scissors, Titanium, Iris, Mortech
Scissors, Titanium, Iris, Mortech
4", Straight
Price
$349.10
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Titanium scissors weigh approximately 25% less than stainless steel, making them the first choice for many technicians. Titanium alloy instruments keep edges sharp longer than traditional instruments. More stain resistant than stainless steel. Helpful in cutting into the facial region.
  • Standard grade titanium stainless steel
Available with straight or curved edges. Do not use bleach or other caustic chemicals.
Specifications
  • Length:
    4"
  • Curvature:
    Straight
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)