Scissors, Titanium, Metzenbaum, Mortech
Product Details & Documents
Titanium alloy scissors weigh approximately 25% less than stainless steel, making them the top choice for many technicians. Titanium alloy instruments are keep edges sharp for longer. More stain resistant than stainless steel. Metzenbaum scissors have a longer handle to blade ratio and can cut extremely delicate tissue.
- Standard grade titanium alloy
- 25% lighter that stainless steel
Available with straight or curved edges. Do not use bleach or other caustic chemicals.
Specifications
- Length:7"
- Curvature:Curved
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.