Orders LinkContactUsLinkComponent
Scissors, Disposable, Mortech
Scissors, Disposable, Mortech
5.25", Sharp/Sharp
Catalog # 76548-606
Supplier:   Mopec
Supplier #:  AA603
Scissors, Disposable, Mortech
Catalog # 76548-606
Supplier:  Mopec
Scissors, Disposable, Mortech
Scissors, Disposable, Mortech
5.25", Sharp/Sharp
Price
$104.64
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
These disposable scissors are highly recommended for post-mortem infectious cases.
  • Straight cutting edge
  • Not autoclavable
  • Single use only
Package of 12. Available in multiple lengths and tips.
Specifications
  • Length:
    5.25"
  • Blade type:
    Sharp/Sharp
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.