Scissors, Disposable, Mortech
Product Details & Documents
These disposable scissors are highly recommended for post-mortem infectious cases.
- Straight cutting edge
- Not autoclavable
- Single use only
Package of 12. Available in multiple lengths and tips.
Specifications
- Length:4.5"
- Blade type:Sharp/Sharp
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)