Scissors, SuperCut Iris, Mortech
Product Details & Documents
SuperCut Scissors are tempered and hardened for a longer, sharper life over ordinary steel. The SuperCut blade has a specially designed sharp end. Also feature rounded and polished finger rings.
- Stainless steel
- Razor sharp cutting edge
Available with straight or curved edges.
Specifications
- Length:4.5"
- Curvature:Curved
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)