Scissors, Superfine Micro, Mortech
Product Details & Documents
Delicate scissors with a fine point for small, very precise, post-mortem dissection.
- Premium grade stainless steel
- Superfine sharp tips
Available with straight or curved edges.
Specifications
- Length:3.25"
- Curvature:Curved
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)