Orders LinkContactUsLinkComponent
Scissors, Iris Dissection, Mortech
Scissors, Iris Dissection, Mortech
4", Straight
Catalog # 76548-484
Supplier:   Mopec
Supplier #:  AA146
Scissors, Iris Dissection, Mortech
Catalog # 76548-484
Supplier:  Mopec
Scissors, Iris Dissection, Mortech
Scissors, Iris Dissection, Mortech
4", Straight
Price
$361.59
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Also referred to as the 'Ribbon Scissor' because of the unique and finger-friendly, comfort handle. With extra wide finger rings offers comfortable post-mortem dissection. This precise and delicate scissor is made of the highest quality German stainless steel. Extra sharp tips allow for intricate work.
  • High grade stainless steel
  • Extra sharp tips
Available with straight or curved tips.
Specifications
  • Length:
    4"
  • Curvature:
    Straight
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.