Orders LinkContactUsLinkComponent
Scissors, Dissection, Sharp/Blunt, Straight, Mortech
Scissors, Dissection, Sharp/Blunt, Straight, Mortech
Premium Grade, 6.5"
Catalog # 76548-418
Supplier:   Mopec
Supplier #:  AA109
Scissors, Dissection, Sharp/Blunt, Straight, Mortech
Catalog # 76548-418
Supplier:  Mopec
Scissors, Dissection, Sharp/Blunt, Straight, Mortech
Scissors, Dissection, Sharp/Blunt, Straight, Mortech
Premium Grade, 6.5"
Price
$231.85
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
These scissors are designed with one sharp tip, one blunt tip and a straight cutting edge. Ideal for a variety of post-mortem dissection needs.
  • Stainless steel
  • Straight cutting edge
  • Sharp/blunt tips
Available in standard or premium grade.
Specifications
  • Description:
    Premium Grade
  • Length:
    6.5"
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)