Scissors, Enterotomy, Mortech
Product Details & Documents
Excellent scissors for small cases.
- Standard grade stainless steel
- Angled end cutting edge
- Lower probe point
Available in right handed and left handed options.
Specifications
- Length:5.5"
- Handle:Right-handed
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)