Scissors, Dissection, Sharp/Blunt, Straight, Mortech
Product Details & Documents
These scissors are designed with one sharp tip, one blunt tip and a straight cutting edge. Ideal for a variety of post-mortem dissection needs.
- Stainless steel
- Straight cutting edge
- Sharp/blunt tips
Available in standard or premium grade.
Specifications
- Description:Standard Grade
- Length:6.5"
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)