Michels Solution , Statlab
Michels Solution, 32 oz
Catalog # 76548-154
Supplier: StatLab
Supplier #: MS-32
Jump to:
Product Details & Documents
Michels Solution is used to preserve and transport specimens for examination in immunofluorescence studies.
- While not a fixative, specimens can be held at room temperature for five days in transport medium before processing
- This simple salt solution maintains pH, but does not kill most pathogens
- Specimens received in transport medium should be washed in three changes of Michels Wash Solution (10 minutes for each wash)
- Not suitable for FISH studies
Part of the StatLab line of formalin alternatives.
Manufactured in the USA
Specifications
Specifications
- Description:Michels Solution
- Size:32 oz