Order Entry
Canada
Orders LinkContactUsLinkComponent
 

Ultra Precision Specific Gravity Hydrometers

1.300...1.370, 330 mm (13"), 0.0005
Catalog # 76546-330
Supplier:   Heathrow Scientific
 :  6602-12
Ultra Precision Specific Gravity Hydrometers
Catalog # 76546-330
Ultra Precision Specific Gravity Hydrometers
Ultra Precision Specific Gravity Hydrometers

1.300...1.370, 330 mm (13"), 0.0005

Price
$51.38
New Customer Promotional Price
Log in for availability
Each
Product Details & Documents

These ultra precision specific gravity hydrometers measure the ratio of the mass of a solid or liquid to the mass of an equal volume of distilled water.

These hydrometers feature smaller ranges but with higher resolution and have all been calibrated for use at 60 °F.

Specifications
  • Length:
    330 mm (13")
  • Range:
    1.300...1.370
  • Subdivisions:
    0.0005