Orders LinkContactUsLinkComponent
Vee Gee Separatory Funnels
Vee Gee Separatory Funnels
1000 ml, 6 (stopcock), 29/42
Catalog # 76546-006
Supplier:   Heathrow Scientific
Supplier #:  1426T-1000
Vee Gee Separatory Funnels
Catalog # 76546-006
Vee Gee Separatory Funnels
Vee Gee Separatory Funnels
1000 ml, 6 (stopcock), 29/42
Price
347,78 $
New Customer Promotional Price
Log in for availability
Each Pack of 4
Product Details & Documents
Our separatory funnels are an excellent tool for liquid-liquid extractions, phase separations, and chemical processes in laboratories.
  • Supplied with PTFE stopcocks to eliminate freezing and lubricant contamination
  • Made from high-quality borosilicate tubing with a low coefficient of expansion
  • Features smooth tooled tips, white block letters, and large marking squares
  • Squibb style
Whether you're in a research laboratory, educational institution, or industrial setting, our separatory funnels empower you to achieve the highest standards of accuracy in your separation procedures.
These separatory funnels feature smooth tooled tips, large white block letters, and large marking squares. Each funnel is made from borosilicate tubing with a low coefficient of expansion, and are supplied with PTFE stopcocks, which eliminates freezing and lubricant contamination.
Specifications
  • Capacity:
    1000 ml
  • [ST] stopper no.:
    29/42
  • Plug size:
    6 (stopcock)