LPX HU TAU CAPTURE BEAD B3 13X 100UL
Catalog # CA76531-118
Supplier: Biolegend
LPX HU TAU CAPTURE BEAD B3 13X 100UL
Catalog # CA76531-118
Supplier: Biolegend
Supplier Number: 741201
Frequently Bought Together
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
PREempt® Concentrate Disinfectants, Contec
Supplier: Contec
pH Test Strips, VWR Chemicals BDH®
Supplier: BDH
Corning® DMEM (Dulbecco's Modification of Eagle's Medium), Corning
Supplier: Corning
Leakbuster™ Specimen Containers
Supplier: Starplex
VWR® Histology Ultra Fine-Tip Pen
Supplier: VWR International
Bright-Line Hemacytometer Set, Hausser Scientific
Supplier: Hausser
VWR® Buffer Solutions
Supplier: VWR
Microscope Immersion Oil, Cargille
Supplier: Cargille Laboratories
Pierce™ Nitrocellulose Membranes, Thermo Scientific
Supplier: Thermo Scientific
Tissue Marking Dye, Flip-Top, Cancer Diagnostics
Supplier: CANCER DIAGNOSTICS, INC.
m Endo LES Agar, for Membrane Filtration, Hardy Diagnostics
Supplier: Hardy Diagnostics
Pierce™ S-Nitrosylation Western Blot Kit, Thermo Scientific
Supplier: Thermo Scientific
Ozone AccuVac® Ampules, MR, Hach
Supplier: Hach