LPX HU CX3CL1 CAPTURE BEAD B5 13X 100UL
Catalog # CA76531-038
Supplier: Biolegend
LPX HU CX3CL1 CAPTURE BEAD B5 13X 100UL
Catalog # CA76531-038
Supplier: Biolegend
Supplier Number: 741169
Frequently Bought Together
VWR® Traceable® Conductivity Calibration Standards (CRM)
Supplier: VWR International
pH Test Strips, VWR Chemicals BDH®
Supplier: BDH
Glycerine ≥99.7% ACS, Laboratory Reagent, VWR Chemicals BDH®
Supplier: VWR International
BD Vacutainer® Venous Blood Collection Tubes
Supplier: BD
Microscope Immersion Oil, Cargille
Supplier: Cargille Laboratories
VWR® Disposable Petri Dishes
Supplier: VWR International
Ethanol denatured, anhydrous
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Single-Use Needles, BD Medical
Supplier: BD
Methanol ≥99.9%, OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
Acetonitrile ≥99.9%, OmniSolv® LC-MS LC-MS for LC-MS, Supelco®
Supplier: MilliporeSigma
Orion™ pH Electrode Cleaning and Storage Solutions, Thermo Scientific
Supplier: Thermo Fisher Scientific
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
Corning® Fetal Bovine Serum, Regular, CA sourced, Corning
Supplier: Corning
Pierce™ Nitrocellulose Membranes, Thermo Scientific
Supplier: Thermo Scientific
MycoAlert™ Mycoplasma Detection Kits, Lonza
Supplier: LONZA PHARMA - BIOSCIENCE CA
Pierce™ S-Nitrosylation Western Blot Kit, Thermo Scientific
Supplier: Thermo Scientific