LPX CCL22 MDC CAPTURE BEAD A6 13X 100UL
Catalog # CA76531-020
Supplier: Biolegend
LPX CCL22 MDC CAPTURE BEAD A6 13X 100UL
Catalog # CA76531-020
Supplier: Biolegend
Supplier Number: 741163
Frequently Bought Together
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
Microscope Immersion Oil, Cargille
Supplier: Cargille Laboratories
Acetone ≥99.5% ACS
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
D-(+)-Trehalose dihydrate ≥99.0%, Reagent Grade
Supplier: VWR
Quality Economy Brand Luer Slip Syringes, Air-Tite
Supplier: Air-Tite
Quality Economy Brand Luer Lock Syringes, Air-Tite
Supplier: Air-Tite
Toluene ≥99.5% ACS, VWR Chemicals BDH®
Supplier: VWR International
Gauze Pads, DUKAL™ Corporation
Supplier: Dukal
UNISETTE™ Processing and Embedding Cassettes, Simport
Supplier: Simport Scientific
Pierce™ Nitrocellulose Membranes, Thermo Scientific
Supplier: Thermo Scientific
High Sensitivity RNA ScreenTape Analysis
Supplier: AGILENT TECHNOLOGIES (GENOMICS) CA
STORAGE SOLUTION PH ELECTRODE
Supplier: Oakton
Pierce™ S-Nitrosylation Western Blot Kit, Thermo Scientific
Supplier: Thermo Scientific
HSW® Norm-Ject® Sterile Luer-Slip Syringes, Air-Tite
Supplier: Air-Tite
Potassium hydrogen carbonate 99%
Supplier: Thermo Scientific Chemicals
DNA MINICOLUMNS V-SPIN HIBIND PK1000
Supplier: Omega Bio-Tek