LPX CCL13 MCP4 CAPTURE BEAD A5 13X 100UL
Catalog # CA76531-018
Supplier: Biolegend
LPX CCL13 MCP4 CAPTURE BEAD A5 13X 100UL
Catalog # CA76531-018
Supplier: Biolegend
Supplier Number: 741162
Frequently Bought Together
Acetone ≥99.5%, HiPerSolv CHROMANORM® for HPLC, VWR Chemicals BDH®
Supplier: VWR International
Potassium chloride 99.0-100.5% ACS, VWR Chemicals BDH®
Supplier: VWR International
Hydrion™, Mikro pH Test Paper with Dispensor, Micro Essential Laboratory
Supplier: Micro Essential
Methanol ≥99.9%, OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
Methanol ≥99.8% ACS, VWR Chemicals BDH®
Supplier: VWR International
Microscope Immersion Oil, Cargille
Supplier: Cargille Laboratories
Tru-Punch™ Sterile Disposable Biopsy Punches, Sklar
Supplier: Sklar
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
Corning® RPMI 1640, Corning
Supplier: Corning
Ozone AccuVac® Ampules, MR, Hach
Supplier: Hach
HSW® Norm-Ject® Sterile Luer-Slip Syringes, Air-Tite
Supplier: Air-Tite
N,N-Dimethylformamide ≥99.8% (by GC), GR ACS, Supelco®
Supplier: MilliporeSigma
Pierce™ Nitrocellulose Membranes, Thermo Scientific
Supplier: Thermo Scientific
Pierce™ S-Nitrosylation Western Blot Kit, Thermo Scientific
Supplier: Thermo Scientific
Acetic acid glacial 99.5-100.5% USP, Multi-Compendial for synthesis, for biochemistry, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Prepared Blood Agar Media
Supplier: Culture Media & Supplies