Order Entry
Canada
Orders LinkContactUsLinkComponent
 

Base Cabinets, Blickman

Countertop, ADA apron assembly with removable front panel, 35"W, C-TOP, —, 35", 75 lbs.
Catalog # 76545-246
Supplier:   BLICKMAN INDUSTRIES
 :  2193500000
Base Cabinets, Blickman
Catalog # 76545-246
Base Cabinets, Blickman
Base Cabinets, Blickman

Countertop, ADA apron assembly with removable front panel, 35"W, C-TOP, —, 35", 75 lbs.

Price
$1,509.94
Log in for availability
Each
 
Product Details & Documents

Stainless steel single and double width base cabinets are constructed with quality 304 stainless steel, #4 polish and have a seamless welded face preventing dirt collection while providing long lasting durability through unparalleled strength and corrosion resistance.

  • Available with 35³/₄" height, 22" depth and 18¹/₈ to 47" width
  • Doors and drawers are equipped with die-cast aluminum bar pulls
  • Offer double walled solid doors for added strength and sound deadening for optimal performance
  • Countertops offered separately

Base cabinets come in different configurations and are designed to stand alone with finished sides or fit within a framework of cabinets.

Specifications
  • Width:
    35"
  • Description:
    Countertop, ADA apron assembly with removable front panel, 35"W, C-TOP
  • Shipping weight:
    75 lbs.
  • Includes: