Order Entry
Canada
Orders LinkContactUsLinkComponent
 

Base Cabinets, Blickman

Double-width stainless steel blickman base cabinet, designed to stand alone with finished sides or to fit within a framework of cabinets, 6 small drawers and 2 large drawers, FC47HS, 47" wide stainless steel base cabinet, standalone or fit within a framework, floor and wall supported, finished sides, four small stainless steel drawers, two (L), two (R), interior dimensions 19⁷/₁₆×2⁵/₈×18¹/₈", two medium stainless steel drawers, one (L), one (R), interior dimensions 19⁷/₁₆×6¼×18⅛", two large stainless steel drawers, interior dimensions 42¹/₄×6¹/₄×18¹/₈", 47", 22", 35¾", 323 lbs.
Catalog # 76545-068
Supplier:   BLICKMAN INDUSTRIES
 :  2013247002
Base Cabinets, Blickman
Catalog # 76545-068
Base Cabinets, Blickman
Base Cabinets, Blickman

Double-width stainless steel blickman base cabinet, designed to stand alone with finished sides or to fit within a framework of cabinets, 6 small drawers and 2 large drawers, FC47HS, 47" wide stainless steel base cabinet, standalone or fit within a framework, floor and wall supported, finished sides, four small stainless steel drawers, two (L), two (R), interior dimensions 19⁷/₁₆×2⁵/₈×18¹/₈", two medium stainless steel drawers, one (L), one (R), interior dimensions 19⁷/₁₆×6¼×18⅛", two large stainless steel drawers, interior dimensions 42¹/₄×6¹/₄×18¹/₈", 47", 22", 35¾", 323 lbs.

Price
$13,641.89
Log in for availability
Each
 
Product Details & Documents

Stainless steel single and double width base cabinets are constructed with quality 304 stainless steel, #4 polish and have a seamless welded face preventing dirt collection while providing long lasting durability through unparalleled strength and corrosion resistance.

  • Available with 35³/₄" height, 22" depth and 18¹/₈ to 47" width
  • Doors and drawers are equipped with die-cast aluminum bar pulls
  • Offer double walled solid doors for added strength and sound deadening for optimal performance
  • Countertops offered separately

Base cabinets come in different configurations and are designed to stand alone with finished sides or fit within a framework of cabinets.

Specifications
  • Depth:
    22"
  • Width:
    47"
  • Description:
    Double-width stainless steel blickman base cabinet, designed to stand alone with finished sides or to fit within a framework of cabinets, 6 small drawers and 2 large drawers, FC47HS
  • Height:
    35¾"
  • Shipping weight:
    323 lbs.
  • Includes:
    47" wide stainless steel base cabinet, standalone or fit within a framework, floor and wall supported, finished sides, four small stainless steel drawers, two (L), two (R), interior dimensions 19⁷/₁₆×2⁵/₈×18¹/₈", two medium stainless steel drawers, one (L), one (R), interior dimensions 19⁷/₁₆×6¼×18⅛", two large stainless steel drawers, interior dimensions 42¹/₄×6¹/₄×18¹/₈"