Order Entry
Canada
Orders LinkContactUsLinkComponent
Base Cabinets, Blickman

Base Cabinets, Blickman

Single-width stainless steel blickman base cabinet, designed to stand alone with finished sides or to fit within a framework of cabinets, four equal size drawers, aE18HS, 18¹/₈" wide stainless steel base cabinet, standalone or fit within a framework, floor and wall supported, finished sides, four stainless steel drawers, interior dimensions (4) 13⁷/₁₆×6¹/₄×18¹/₈", 18⅛", 22", 35¾", 174 lbs.
  76545-050
Supplier #:  2012318000
Base Cabinets, Blickman
  76545-050
Base Cabinets, Blickman
Base Cabinets, Blickman

Single-width stainless steel blickman base cabinet, designed to stand alone with finished sides or to fit within a framework of cabinets, four equal size drawers, aE18HS, 18¹/₈" wide stainless steel base cabinet, standalone or fit within a framework, floor and wall supported, finished sides, four stainless steel drawers, interior dimensions (4) 13⁷/₁₆×6¹/₄×18¹/₈", 18⅛", 22", 35¾", 174 lbs.

 
$6,919.80
   
Each
Minimum order quantity is 1
 

Stainless steel single and double width base cabinets are constructed with quality 304 stainless steel, #4 polish and have a seamless welded face preventing dirt collection while providing long lasting durability through unparalleled strength and corrosion resistance.

  • Available with 35³/₄" height, 22" depth and 18¹/₈ to 47" width
  • Doors and drawers are equipped with die-cast aluminum bar pulls
  • Offer double walled solid doors for added strength and sound deadening for optimal performance
  • Countertops offered separately

Base cabinets come in different configurations and are designed to stand alone with finished sides or fit within a framework of cabinets.

Documents

 
  • Depth:
    22"
  • Width:
    18⅛"
  • Description:
    Single-width stainless steel blickman base cabinet, designed to stand alone with finished sides or to fit within a framework of cabinets, four equal size drawers, aE18HS
  • Height:
    35¾"
  • Shipping weight:
    174 lbs.
  • Includes:
    18¹/₈" wide stainless steel base cabinet, standalone or fit within a framework, floor and wall supported, finished sides, four stainless steel drawers, interior dimensions (4) 13⁷/₁₆×6¹/₄×18¹/₈"