Order Entry
Canada
Orders LinkContactUsLinkComponent
Base Cabinets, Blickman | Avantor

Base Cabinets, Blickman

Double-width stainless steel blickman base cabinet, designed to stand alone with finished sides or to fit within a framework of cabinets, 2 small drawers and 3 large drawers, DS35HS, 35" wide stainless steel base cabinet, standalone or fit within a framework, floor and wall supported, finished sides, five stainless steel drawers, interior dimensions (2) 30¹/₄×2⁵/₈×18¹/₈" over (3) 30¹/₄×6¹/₄×18¹/₈", 35", 22", 35¾", 275 lbs.
Catalog # 76544-996
Supplier:   BLICKMAN INDUSTRIES
Fournisseur #:  2013735001
Base Cabinets, Blickman
Catalog # 76544-996
Base Cabinets, Blickman
Base Cabinets, Blickman

Double-width stainless steel blickman base cabinet, designed to stand alone with finished sides or to fit within a framework of cabinets, 2 small drawers and 3 large drawers, DS35HS, 35" wide stainless steel base cabinet, standalone or fit within a framework, floor and wall supported, finished sides, five stainless steel drawers, interior dimensions (2) 30¹/₄×2⁵/₈×18¹/₈" over (3) 30¹/₄×6¹/₄×18¹/₈", 35", 22", 35¾", 275 lbs.

Price
$9,843.36
Log in for availability
Each
La quantité minimale de commande est de 1
Product Details & Documents

Stainless steel single and double width base cabinets are constructed with quality 304 stainless steel, #4 polish and have a seamless welded face preventing dirt collection while providing long lasting durability through unparalleled strength and corrosion resistance.

  • Available with 35³/₄" height, 22" depth and 18¹/₈ to 47" width
  • Doors and drawers are equipped with die-cast aluminum bar pulls
  • Offer double walled solid doors for added strength and sound deadening for optimal performance
  • Countertops offered separately

Base cabinets come in different configurations and are designed to stand alone with finished sides or fit within a framework of cabinets.

Documents

Specifications
  • Depth:
    22"
  • Width:
    35"
  • Description:
    Double-width stainless steel blickman base cabinet, designed to stand alone with finished sides or to fit within a framework of cabinets, 2 small drawers and 3 large drawers, DS35HS
  • Height:
    35¾"
  • Shipping weight:
    275 lbs.
  • Includes:
    35" wide stainless steel base cabinet, standalone or fit within a framework, floor and wall supported, finished sides, five stainless steel drawers, interior dimensions (2) 30¹/₄×2⁵/₈×18¹/₈" over (3) 30¹/₄×6¹/₄×18¹/₈"
Recommandations

Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".

Sinon, vous recevrez des recommandations génériques.