Order Entry
Canada
Orders LinkContactUsLinkComponent
 

Base Cabinets, Blickman

Single-width stainless steel blickman base cabinet, designed to stand alone with finished sides or to fit within a framework of cabinets, one compartment, aA24HS, 24¹/₈" wide stainless steel base cabinet, standalone or fit within a framework, floor and wall supported, finished sides, one compartment that includes one stainless steel shelf inside, 24⅛", 22", 35¾", 132 lbs.
Catalog # 76544-974
Supplier:   BLICKMAN INDUSTRIES
 :  2012224000
Base Cabinets, Blickman
Catalog # 76544-974
Base Cabinets, Blickman
Base Cabinets, Blickman

Single-width stainless steel blickman base cabinet, designed to stand alone with finished sides or to fit within a framework of cabinets, one compartment, aA24HS, 24¹/₈" wide stainless steel base cabinet, standalone or fit within a framework, floor and wall supported, finished sides, one compartment that includes one stainless steel shelf inside, 24⅛", 22", 35¾", 132 lbs.

Price
4 663,07 $
Log in for availability
Each
 
Product Details & Documents

Stainless steel single and double width base cabinets are constructed with quality 304 stainless steel, #4 polish and have a seamless welded face preventing dirt collection while providing long lasting durability through unparalleled strength and corrosion resistance.

  • Available with 35³/₄" height, 22" depth and 18¹/₈ to 47" width
  • Doors and drawers are equipped with die-cast aluminum bar pulls
  • Offer double walled solid doors for added strength and sound deadening for optimal performance
  • Countertops offered separately

Base cabinets come in different configurations and are designed to stand alone with finished sides or fit within a framework of cabinets.

Specifications
  • Depth:
    22"
  • Width:
    24⅛"
  • Description:
    Single-width stainless steel blickman base cabinet, designed to stand alone with finished sides or to fit within a framework of cabinets, one compartment, aA24HS
  • Height:
    35¾"
  • Shipping weight:
    132 lbs.
  • Includes:
    24¹/₈" wide stainless steel base cabinet, standalone or fit within a framework, floor and wall supported, finished sides, one compartment that includes one stainless steel shelf inside